Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zfp68
Entrez Gene ID
304337
Gene Name
zinc finger protein 68
Gene Aliases
-
Chromosome Location
Chr.12: 18152595 - 18167523 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107128.2 NP_001100598.2 4 - 5 419 110
NM_001142945.1 NP_001136417.1 3 - 4 323 110
XM_008768974.1 XP_008767196.1 1 - 2 788 110
XM_008768975.1 XP_008767197.1 4 - 5 477 110

View Full Product Details