Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfp68
Entrez Gene ID
304337
Gene Name
zinc finger protein 68
Gene Aliases
-
Chromosome Location
Chr.12: 18152595 - 18167523 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107128.2 NP_001100598.2 5 - 6 505 80
NM_001142945.1 NP_001136417.1 4 - 5 409 80
XM_008768974.1 XP_008767196.1 2 - 3 874 80
XM_008768975.1 XP_008767197.1 5 - 6 563 80

View Full Product Details