Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mmd2
Entrez Gene ID
304301
Gene Name
monocyte to macrophage differentiation-associated 2
Gene Aliases
-
Chromosome Location
Chr.12: 14021724 - 14078121 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037217.1 NP_001032294.1 3 - 4 462 84
GenBank mRNA BC161816.1 - 3 - 4 440 84
CO400908.1 - 3 - 4 439 84

View Full Product Details