Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lfng
Entrez Gene ID
170905
Gene Name
LFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
Gene Aliases
-
Chromosome Location
Chr.12: 16118192 - 16126211 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133393.2 NP_596884.1 3 - 4 638 133
GenBank mRNA AB054539.1 - 3 - 4 579 133
BC070933.1 - 3 - 4 638 133

View Full Product Details