Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zcchc12
Entrez Gene ID
313436
Gene Name
zinc finger, CCHC domain containing 12
Gene Aliases
-
Chromosome Location
Chr.X: 122808605 - 122811790 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014065.2 NP_001014087.1 4 - 5 276 78
XM_006257448.2 XP_006257510.1 4 - 5 275 78
XM_006257449.2 XP_006257511.1 4 - 5 286 78
XM_006257451.2 XP_006257513.1 4 - 5 327 78
XM_006257452.2 XP_006257514.1 3 - 4 258 78
XM_008773415.1 XP_008771637.1 3 - 4 262 78
GenBank mRNA BC089115.1 - 4 - 5 276 78

View Full Product Details