Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zc3h4
Entrez Gene ID
678741
Gene Name
zinc finger CCCH-type containing 4
Gene Aliases
-
Chromosome Location
Chr.1: 78352810 - 78390906 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001053214.4 XP_001053214.2 5 - 6 841 104
XM_006228482.1 XP_006228544.1 5 - 6 1013 104
XM_006228483.2 XP_006228545.1 6 - 7 1550 104
XM_006228484.2 XP_006228546.1 6 - 7 904 104
XM_006228485.2 XP_006228547.1 6 - 7 1233 104
XM_006228486.2 XP_006228548.1 5 - 6 1405 104
XM_006228487.1 XP_006228549.1 5 - 6 764 104
XM_006228488.2 XP_006228550.1 6 - 7 1116 104
XM_006228489.1 XP_006228551.1 6 - 7 903 104
XM_006228491.2 XP_006228553.1 6 - 7 1027 104
XM_006228493.1 XP_006228555.1 5 - 6 1013 104
XM_008759031.1 XP_008757253.1 5 - 6 958 104

View Full Product Details