Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zc3h4
Entrez Gene ID
678741
Gene Name
zinc finger CCCH-type containing 4
Gene Aliases
-
Chromosome Location
Chr.1: 78352810 - 78390906 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006228493.1 XP_006228555.1 12 - 13 2318 60

View Full Product Details