Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bcl3
Entrez Gene ID
680611
Gene Name
B-cell CLL/lymphoma 3
Gene Aliases
-
Chromosome Location
Chr.1: 80730758 - 80745086 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109422.1 NP_001102892.1 8 - 9 1200 150

View Full Product Details