Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pdcd2l
Entrez Gene ID
689637
Gene Name
programmed cell death 2-like
Gene Aliases
RGD1308249
Chromosome Location
Chr.1: 90045468 - 90057756 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109544.1 NP_001103014.1 1 - 2 140 82

View Full Product Details