Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Igsf23
Entrez Gene ID
502306
Gene Name
immunoglobulin superfamily, member 23
Gene Aliases
RGD1564357
Chromosome Location
Chr.1: 80850091 - 80865391 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006228504.2 XP_006228566.1 5 - 6 1160 69

View Full Product Details