Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sphk2
Entrez Gene ID
308589
Gene Name
sphingosine kinase 2
Gene Aliases
-
Chromosome Location
Chr.1: 101692961 - 101700604 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012066.1 NP_001012066.1 3 - 4 653 82
XM_006229085.1 XP_006229147.1 3 - 4 742 82
XM_006229086.2 XP_006229148.1 3 - 4 1030 82
XM_006229088.2 XP_006229150.1 3 - 4 802 82
XM_008759383.1 XP_008757605.1 3 - 4 695 82
XM_008759384.1 XP_008757606.1 3 - 4 1084 82
XM_008759385.1 XP_008757607.1 3 - 4 1037 82
GenBank mRNA BC079120.1 - 3 - 4 653 82
FQ211663.1 - 3 - 4 819 82

View Full Product Details