Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Psg29
Entrez Gene ID
292666
Gene Name
pregnancy-specific glycoprotein 29
Gene Aliases
-
Chromosome Location
Chr.1: 79236391 - 79259492 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025641.1 NP_001020812.1 2 - 3 531 94
GenBank mRNA BC097354.1 - 2 - 3 531 94

View Full Product Details