The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001082549.1 | NP_001076018.1 | 6 - 7 | 874 | 116 |
NM_199087.1 | NP_954518.1 | 5 - 6 | 703 | 116 | |
GenBank mRNA | BC061768.1 | - | 5 - 6 | 703 | 116 |