Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rbm42
Entrez Gene ID
361545
Gene Name
RNA binding motif protein 42
Gene Aliases
RGD1306184
Chromosome Location
Chr.1: 89099296 - 89109251 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014159.2 NP_001014181.2 2 - 3 372 117
XM_006228781.2 XP_006228843.1 2 - 3 397 117
GenBank mRNA BC079321.1 - 2 - 3 330 117
FQ222852.1 - 2 - 3 342 117

View Full Product Details