The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014159.2 | NP_001014181.2 | 8 - 9 | 1229 | 102 |
XM_006228781.2 | XP_006228843.1 | 7 - 8 | 1167 | 102 | |
GenBank mRNA | BC079321.1 | - | 8 - 9 | 1187 | 102 |
FQ216075.1 | - | 4 - 5 | 611 | 102 | |
FQ222852.1 | - | 7 - 8 | 1112 | 102 | |
FQ235070.1 | - | 4 - 5 | 579 | 102 |