The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012063.1 | NP_001012063.1 | 8 - 9 | 999 | 67 |
GenBank mRNA | BC079411.1 | - | 8 - 9 | 999 | 67 |