The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106260.1 | NP_001099730.1 | 7 - 8 | 775 | 90 |
XM_006229025.1 | XP_006229087.1 | 7 - 8 | 928 | 90 | |
XM_006229026.2 | XP_006229088.1 | 8 - 9 | 960 | 90 | |
GenBank mRNA | FQ216351.1 | - | 7 - 8 | 895 | 90 |