The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110491.1 | NP_001103961.1 | - | 491 | 59 |
GenBank mRNA | BC091204.1 | - | - | 491 | 59 |