Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxo27
Entrez Gene ID
499114
Gene Name
F-box protein 27
Gene Aliases
RGD1563982
Chromosome Location
Chr.1: 86872808 - 86878709 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110491.1 NP_001103961.1 - 700 80
GenBank mRNA BC091204.1 - - 700 80

View Full Product Details