Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Qpctl
Entrez Gene ID
292687
Gene Name
glutaminyl-peptide cyclotransferase-like
Gene Aliases
RGD1308128
Chromosome Location
Chr.1: 80047504 - 80057016 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106230.1 NP_001099700.1 3 - 4 695 66
XM_006228402.2 XP_006228464.1 3 - 4 1137 66
GenBank mRNA BC169076.1 - 3 - 4 691 66

View Full Product Details