Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Psip1
Entrez Gene ID
313323
Gene Name
PC4 and SFRS1 interacting protein 1
Gene Aliases
Ledgf, Psip2, SBP75
Chromosome Location
Chr.5: 101556198 - 101588664 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175765.2 NP_786941.1 11 - 12 1196 137
XM_006238353.2 XP_006238415.1 12 - 13 1423 137
XM_006238354.2 XP_006238416.1 12 - 13 1419 137
GenBank mRNA AB285525.1 - - 1099 137
AF339084.1 - 11 - 12 1196 137

View Full Product Details