Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rbm11
Entrez Gene ID
288321
Gene Name
RNA binding motif protein 11
Gene Aliases
-
Chromosome Location
Chr.11: 14005701 - 14015723 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105898.1 NP_001099368.1 4 - 5 499 153

View Full Product Details