The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008287.1 | NP_001008288.1 | 3 - 4 | 72 | |
XM_006248047.2 | XP_006248109.1 | 3 - 4 | 689 | 72 | |
XM_006248048.2 | XP_006248110.1 | 3 - 4 | 689 | 72 | |
GenBank mRNA | BC085847.1 | - | 3 - 4 | 72 |