Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Donson
Entrez Gene ID
288257
Gene Name
downstream neighbor of SON
Gene Aliases
-
Chromosome Location
Chr.11: 31834644 - 31847490 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006248047.2 XP_006248109.1 9 - 10 1650 66
XM_006248048.2 XP_006248110.1 9 - 10 1650 66

View Full Product Details