Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lmna
Entrez Gene ID
60374
Gene Name
lamin A/C
Gene Aliases
-
Chromosome Location
Chr.2: 187842884 - 187863552 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002016.2 NP_001002016.2 7 - 8 1588 95
GenBank mRNA BC062018.1 - 6 - 7 1143 95
CA338388.1 - 3 - 4 573 95
FM060776.1 - 1 - 2 125 95
X66870.1 - 7 - 8 1547 95
X76297.1 - 7 - 8 1310 95
X99257.1 - 7 - 8 1047 95

View Full Product Details