Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fxyd4
Entrez Gene ID
64190
Gene Name
FXYD domain-containing ion transport regulator 4
Gene Aliases
Chif
Chromosome Location
Chr.4: 149984206 - 149988743 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022388.1 NP_071783.1 4 - 5 1018 76
XM_006237206.2 XP_006237268.1 5 - 6 357 76
XM_008763260.1 XP_008761482.1 5 - 6 687 76
XM_008763261.1 XP_008761483.1 5 - 6 645 76
GenBank mRNA L41254.1 - 4 - 5 1018 76

View Full Product Details