The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012705.1 | NP_036837.1 | 8 - 9 | 1397 | 135 |
XM_006237331.2 | XP_006237393.1 | 10 - 11 | 1660 | 135 | |
XM_006237333.2 | XP_006237395.1 | 8 - 9 | 1420 | 135 | |
XM_008763280.1 | XP_008761502.1 | 8 - 9 | 1377 | 135 | |
GenBank mRNA | M15768.1 | - | 8 - 9 | 1397 | 135 |