Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cops7a
Entrez Gene ID
312710
Gene Name
COP9 signalosome subunit 7A
Gene Aliases
-
Chromosome Location
Chr.4: 157479549 - 157487152 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047098.2 NP_001040563.2 3 - 4 388 103
NM_001164091.1 NP_001157563.1 3 - 4 404 103
XM_006237367.1 XP_006237429.1 3 - 4 412 103
XM_006237368.2 XP_006237430.1 3 - 4 598 103
GenBank mRNA AB259153.1 - 2 - 3 116 103
FM059742.1 - 3 - 4 388 103
FM122308.1 - 3 - 4 404 103
FQ215513.1 - 3 - 4 412 103

View Full Product Details