Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cops7a
Entrez Gene ID
312710
Gene Name
COP9 signalosome subunit 7A
Gene Aliases
-
Chromosome Location
Chr.4: 157479549 - 157487152 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047098.2 NP_001040563.2 4 - 5 472 123
NM_001164091.1 NP_001157563.1 4 - 5 488 123
XM_006237367.1 XP_006237429.1 4 - 5 496 123
XM_006237368.2 XP_006237430.1 4 - 5 682 123
GenBank mRNA AB259153.1 - 3 - 4 200 123
FM059742.1 - 4 - 5 472 123
FM122308.1 - 4 - 5 488 123
FQ215513.1 - 4 - 5 496 123

View Full Product Details