Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Foxj2
Entrez Gene ID
502886
Gene Name
forkhead box J2
Gene Aliases
RGD1565101
Chromosome Location
Chr.4: 155653718 - 155679815 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109352.1 NP_001102822.1 5 - 6 1405 155
XM_006237311.2 XP_006237373.1 6 - 7 1218 155

View Full Product Details