Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tas2r13
Entrez Gene ID
78984
Gene Name
taste receptor, type 2, member 13
Gene Aliases
T2R8
Chromosome Location
Chr.4: 166958466 - 166959410 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023998.1 NP_076488.1 1 - 1 258 122
GenBank mRNA AF227145.1 - 1 - 1 258 122

View Full Product Details