Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vgll4
Entrez Gene ID
297523
Gene Name
vestigial-like family member 4
Gene Aliases
-
Chromosome Location
Chr.4: 146777756 - 146890050 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008763229.1 XP_008761451.1 1 - 2 262 63

View Full Product Details