Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Magohb
Entrez Gene ID
690303
Gene Name
mago-nashi homolog B (Drosophila)
Gene Aliases
-
Chromosome Location
Chr.4: 165514582 - 165522366 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191105.1 NP_001178034.1 1 - 2 99 110

View Full Product Details