Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc42
Entrez Gene ID
303234
Gene Name
coiled-coil domain containing 42
Gene Aliases
RGD1565925
Chromosome Location
Chr.10: 55169312 - 55179489 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107009.1 NP_001100479.1 6 - 7 1040 99

View Full Product Details