Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr81
Entrez Gene ID
303312
Gene Name
WD repeat domain 81
Gene Aliases
RGD1311334
Chromosome Location
Chr.10: 62273817 - 62287207 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134360.1 NP_001127832.1 4 - 5 4340 82

View Full Product Details