Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rhbdf1
Entrez Gene ID
303008
Gene Name
rhomboid 5 homolog 1 (Drosophila)
Gene Aliases
-
Chromosome Location
Chr.10: 15672382 - 15685285 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030034.1 NP_001025205.1 17 - 18 2286 91
XM_006245977.2 XP_006246039.1 17 - 18 6104 91
XM_008767578.1 XP_008765800.1 18 - 19 5221 91
XM_008767579.1 XP_008765801.1 18 - 19 5096 91
XM_008767580.1 XP_008765802.1 17 - 18 3206 91
GenBank mRNA BC099777.1 - 17 - 18 2286 91

View Full Product Details