Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc16a6
Entrez Gene ID
303772
Gene Name
solute carrier family 16, member 6
Gene Aliases
Mct7B
Chromosome Location
Chr.10: 97733701 - 97758734 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006247620.2 XP_006247682.1 1 - 2 1952 100

View Full Product Details