Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Luc7l3
Entrez Gene ID
360602
Gene Name
LUC7-like 3 (S. cerevisiae)
Gene Aliases
Crop, RGD1307981
Chromosome Location
Chr.10: 81991147 - 82027119 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108291.1 NP_001101761.1 3 - 4 367 85
XM_006247155.2 XP_006247217.1 3 - 4 359 85
XM_006247156.2 XP_006247218.1 3 - 4 358 85
XM_008768078.1 XP_008766300.1 3 - 4 358 85
GenBank mRNA FQ235074.1 - 3 - 4 333 85

View Full Product Details