Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cwc25
Entrez Gene ID
360613
Gene Name
CWC25 spliceosome-associated protein homolog (S. cerevisiae)
Gene Aliases
Ccdc49, RGD1305258
Chromosome Location
Chr.10: 85684898 - 85709568 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108295.1 NP_001101765.1 7 - 8 992 84
GenBank mRNA BC168194.1 - 5 - 6 700 84

View Full Product Details