Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tlcd1
Entrez Gene ID
287472
Gene Name
TLC domain containing 1
Gene Aliases
-
Chromosome Location
Chr.10: 65439104 - 65441140 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008767957.1 XP_008766179.1 1 - 2 483 98

View Full Product Details