Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
G3bp1
Entrez Gene ID
171092
Gene Name
GTPase activating protein (SH3 domain) binding protein 1
Gene Aliases
G3bp, RGD1310666
Chromosome Location
Chr.10: 40812858 - 40846252 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133565.1 NP_598249.1 8 - 9 924 82
GenBank mRNA FQ217639.1 - 7 - 8 2925 82
FQ217678.1 - 7 - 8 2924 82

View Full Product Details