The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012977.1 | NP_037109.1 | 7 - 8 | 646 | 81 |
GenBank mRNA | BC061566.1 | - | 7 - 8 | 607 | 81 |
U59462.1 | - | 7 - 8 | 568 | 81 | |
U67958.1 | - | 7 - 8 | 646 | 81 | |
U72741.1 | - | 8 - 9 | 664 | 81 |