Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Abca8
Entrez Gene ID
303637
Gene Name
ATP-binding cassette, subfamily A (ABC1), member 8
Gene Aliases
Abca8b
Chromosome Location
Chr.10: 98236919 - 98308290 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008768450.1 XP_008766672.1 28 - 29 4280 58

View Full Product Details