Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Kctd11
- Entrez Gene ID
-
363634
- Gene Name
- potassium channel tetramerization domain containing 11
- Gene Aliases
-
-
- Chromosome Location
-
Chr.10: 56489552 - 56491715 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC168958.1 |
- |
- |
873 |
71 |
- Gene Symbol
- Tmem95
- Entrez Gene ID
-
691982
- Gene Name
- transmembrane protein 95
- Gene Aliases
-
-
- Chromosome Location
-
Chr.10: 56486765 - 56488882 on Build Rnor_6.0
View Full Product Details