Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Csf3
Entrez Gene ID
25610
Gene Name
colony stimulating factor 3 (granulocyte)
Gene Aliases
Gcsf
Chromosome Location
Chr.10: 86616708 - 86619160 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017104.1 NP_058800.1 2 - 3 214 63
XM_006247265.2 XP_006247327.2 2 - 3 283 63
GenBank mRNA U37101.1 - 2 - 3 214 63

View Full Product Details