Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rasgef1c
Entrez Gene ID
360519
Gene Name
RasGEF domain family, member 1C
Gene Aliases
-
Chromosome Location
Chr.10: 35392762 - 35459267 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108273.1 NP_001101743.1 5 - 6 762 79

View Full Product Details