Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Scrn2
Entrez Gene ID
360612
Gene Name
secernin 2
Gene Aliases
-
Chromosome Location
Chr.10: 84961439 - 84971022 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012142.1 NP_001012142.1 5 - 6 824 76
XM_006247342.2 XP_006247404.1 5 - 6 1053 76
XM_008768079.1 XP_008766301.1 5 - 6 2337 76
GenBank mRNA BC078939.1 - 5 - 6 824 76

View Full Product Details