The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039033.1 | NP_001034122.1 | 12 - 13 | 1198 | 106 |
XM_006232253.2 | XP_006232315.1 | 12 - 13 | 1226 | 106 | |
XM_008760930.1 | XP_008759152.1 | 12 - 13 | 1266 | 106 | |
XM_008760931.1 | XP_008759153.1 | 12 - 13 | 1242 | 106 | |
GenBank mRNA | BC098698.1 | - | 12 - 13 | 1198 | 106 |