Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cpa3
Entrez Gene ID
54242
Gene Name
carboxypeptidase A3, mast cell
Gene Aliases
-
Chromosome Location
Chr.2: 105016798 - 105047989 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019300.1 NP_062173.1 6 - 7 578 119
GenBank mRNA U67914.1 - 6 - 7 564 119

View Full Product Details