Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Upf3b
Entrez Gene ID
313449
Gene Name
UPF3 regulator of nonsense transcripts homolog B (yeast)
Gene Aliases
RGD1560264
Chromosome Location
Chr.X: 123713327 - 123731426 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135873.1 NP_001129345.1 9 - 10 1380 69
XM_006257455.2 XP_006257517.1 10 - 11 1442 69
GenBank mRNA FQ234598.1 - 2 - 3 396 69

View Full Product Details